missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBL Antibody (OTI1A12), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72594V
This item is not returnable.
View return policy
Description
MBL Monoclonal specifically detects MBL in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MBL | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MBL2 | |
| Full length recombinant protein of human MBL (NP_000233) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1A12 | |
| Western Blot, Immunohistochemistry | |
| COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBLmannan-binding lectin, MBP, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833 | |
| Mouse | |
| Affinity Purified | |
| Immunology | |
| 4153.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction