missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MBL1 Monoclonal Antibody (8G6)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA533352

Product Code. 17105253

  • € 1066.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product is supplied at 1 mL (100 μL/mL). Application Notes: Western Blot - A non-reduced sample treatment was used. The band sizes are 191, 263 and 316 kDa. IHC-F - Tissue sections were fixed in 4% PBS-buffered formaldehyde and pretreated with 2% hydrogen peroxide in methanol for 20 minutes at 4°C to quench endogenous peroxidases. Rat IgG2a was used as a negative control.

Regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. Necessary for centrosome duplication. It functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MBL1
Monoclonal
100 μg/mL
PBS with 0.1% BSA and 0.02% sodium azide
Q9Z2D8
MBD3
Purified mouse MBL-A.
100 μg
Primary
Mouse
Antibody
IgG2a
ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
8G6
Unconjugated
MBD3
AI181826; AU019209; CpG binding domain 3; Mbd3; methyl-CpG binding domain protein 3; methyl-CpG-binding domain protein 3; Methyl-CpG-binding protein MBD3
Rat
Protein G
RUO
17192
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.