missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Antibody (VEGF/1063), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11437H
This item is not returnable.
View return policy
Description
VEGF Monoclonal antibody specifically detects VEGF in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| VEGF | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| VEGF/1063 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Human VEGF recombinant protein (Uniprot: P15692) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 7422 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction