missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MBNL1 Monoclonal Antibody (1H1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547391

 View more versions of this product

Product Code. 17845267

  • € 504.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MBNL1 Monoclonal Antibody for Western Blot, ICC/IF

EGFR (Epidermal growth factor receptor, HER1, ErbB1) is encoded by the EGFR gene located on chromosome 7 in humans. EGFR belongs to the HER/ERbB family of proteins that includes three other receptor tyrosine kinases, ERbB2, ERbB3, ERbB4. EGFR is a transmembrane receptor and binding of its cognate ligands such as EGF (Epidermal Growth Factor) and TGF alpha (Transforming Growth Factor alpha) to the extracellular domain leads to EGFR dimerization followed by autophosphorylation of the tyrosine residues in the cytoplasmic domain. Phosphorylation of EGFR at certain residues is also mediated by Src-non-receptor kinase. EGFR activation signals multiple downstream signaling cascades such as the Ras - ERK, PI3-K - Akt, Jak - STAT and PKC pathways that help in growth and proliferation of cells. Phosphorylation of EGFR at Y1086 specifically allows binding of the adaptor protein GRB2, leading to activation of the MAPK pathway. Upon receptor activation and signaling, EGFR is endocytosed and targeted for degradation or recycling. Mutations in the EGFR gene are associated with lung cancer and multiple alternatively spliced transcript variants encode different protein isoforms of EGFR have been found. Increased production or activation of EGFR has been associated with poor prognosis in a variety of tumors. Moreover, EGFR overexpression is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MBNL1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
Q9NR56
MBNL1
Full-length recombinant human muscleblind-like 1 protein expressed in and purified from E. coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
1H1
Unconjugated
MBNL1
DKFZp686P06174; EXP; EXP35; EXP40; EXP42; KIAA0428; MBNL; MBNL1; mKIAA0428; muscleblind like splicing regulator 1; muscleblind-like 1; muscleblind-like 1 (Drosophila); muscleblind-like protein 1; muscleblind-like splicing regulator 1; triplet-expansion RNA-binding protein
Mouse
Protein G
RUO
4154
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.