missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBNL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-43638
This item is not returnable.
View return policy
Description
Specifically detects MBNL2 in Human, Mouse samples, and it is validated for Immunohistochemistry (Paraffin)
MBNL2 Polyclonal specifically detects MBNL2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MBNL2 | |
| Polyclonal | |
| Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| DKFZp781H1296, MBLL39Muscleblind-like protein 1, MBLLMGC120628, MGC120625, MGC120626, MLP1, muscleblind-like 2 (Drosophila), muscleblind-like protein 2, Muscleblind-like protein-like, Muscleblind-like protein-like 39, PRO2032 | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| MBNL2 | |
| Recombinant protein encompassing a sequence within the center region of human MBNL2. The exact sequence is proprietary. | |
| Immunogen affinity purified | |
| RUO | |
| 10150 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction