missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBNL3 Antibody (PCRP-MBNL3-1D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08663CL1
This item is not returnable.
View return policy
Description
MBNL3 Monoclonal antibody specifically detects MBNL3 in Human samples. It is validated for Flow Cytometry,Protein Array
Specifications
| MBNL3 | |
| Monoclonal | |
| CoraFluor 1 | |
| CHCRMBLX39FLJ11316, Cys3His CCG1-required protein, FLJ97142, MBXLMBLX, muscleblind-like 3 (Drosophila), muscleblind-like protein 3, Muscleblind-like X-linked protein, Protein HCHCR | |
| Recombinant full-length human MBNL3 protein (Uniprot: Q9NUK0) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Protein Array | |
| PCRP-MBNL3-1D11 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 55796 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction