missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBOAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543193
This item is not returnable.
View return policy
Description
Peptide sequence: AAEPQPSSLS YRTTGSTYLH PLSELLGIPL DQVNFVVCQL VALFAAFWFR Sequence homology: Horse: 100%; Human: 100%; Mouse: 91%; Pig: 93%; Rabbit: 86%; Rat: 91%.
MBOAT1 shares structural similarity with a superfamily of membrane-bound O-acetyltransferases that transfer organic compounds, usually fatty acids (e.g., cholesterol, diacylglycerol, palmitoyl), onto hydroxyl groups of membrane-embedded targets.MBOAT1 shares structural similarity with a superfamily of membrane-bound O-acetyltransferases that transfer organic compounds, usually fatty acids (e.g., cholesterol, diacylglycerol, palmitoyl), onto hydroxyl groups of membrane-embedded targets (Dauwerse et al., 2007 [PubMed 17440500]).[supplied by OMIM]. Sequence Note: The RefSeq transcript and protein were derived from genomic sequence to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments.
Specifications
| MBOAT1 | |
| Polyclonal | |
| Unconjugated | |
| Mboat1 | |
| 1-acylglycerophosphoethanolamine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase; 9130215M02Rik; BC023845; dJ434O11.1; LPE acyltransferase 1; LPEAT; Lpeat1; LPLAT; LPLAT 1; LPSAT; lyso-PE acyltransferase; lysophosphatidylethanolamine acyltransferase; lysophosphatidylethanolamine acyltransferase 1; lysophosphatidylserine acyltransferase; Lysophospholipid acyltransferase 1; lyso-PS acyltransferase; MBOAT1; membrane bound O-acyl transferase family, member 1; membrane bound O-acyltransferase domain containing 1; membrane-bound O-acyltransferase domain-containing protein 1; MGC44669; Moact1; OACT1; O-acyltransferase (membrane bound) domain containing 1; O-acyltransferase domain-containing protein 1; RGD1565521; RGD1565561 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 154141 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6ZNC8 | |
| Mboat1 | |
| Synthetic peptide directed towards the N-terminal of human MBOAT1 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction