missing translation for 'onlineSavingsMsg'
Learn More

MBP Antibody (12), Novus Biologicals™

Rat Monoclonal Antibody has been used in 19 publications

€ 207.00

Specifications

Antigen MBP
Clone 12
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunodiffusion
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18666451
View Documents
Novus Biologicals
NB600-717SS
0.25 mL
€ 207.00
250µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MBP Monoclonal specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Mammal, Rabbit, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunodiffusion.
Specifications

Specifications

MBP
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunodiffusion
Monoclonal
Supernatant
RUO
Human
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
MBP
IgG2a
Tissue culture supernatant
NB600-717 reacts with myelin basic protein from a wide range of species. The antibody reacts weakly with peptides ending in the Phe 91 where the 91-92 Phe-Phe bond is broken. Synthetic peptide 82-99 reacts very well, as does intact MBP. Further epitope analysis indicates binding to a region defined by amino acids 82-87 (DENPVV). Clone 12 has been reported as being suitable for use in Western blotting.
12
Western Blot
Unconjugated
Rat
Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
P02687
4155
Bovine MBP
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.