missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (2H9), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22121PCP
This item is not returnable.
View return policy
Description
MBP Monoclonal specifically detects MBP in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| MBP | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| MBP | |
| Purified recombinant fragment of human Myelin Basic Protein expressed in E. coli. [UniProt# P02686] | |
| 0.1 mL | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4155 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2H9 | |
| Flow Cytometry | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction