missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (CL2819) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46630JF646
This item is not returnable.
View return policy
Description
MBP Monoclonal antibody specifically detects MBP in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| MBP | |
| Monoclonal | |
| Janelia Fluor 646 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| This antibody was developed using a recombinant protein derived from P02686, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4155 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL2819 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur