missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-79873FR
This item is not returnable.
View return policy
Description
MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry - Free Floating
Specifications
| MBP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Reacts with residues 49-62 of Isoforms 5 and 6 of the human protein. The immunogen corresponds to Isoform 1 between amino acids 182-195 of the 33 kDa isoform. The antibody also detects mouse and rat MBP protein. The antibody primarily detects a 22 kDa band. | |
| 0.1 mL | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
| 4155 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction