missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (MBP/4277R), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08388AF350
This item is not returnable.
View return policy
Description
MBP Monoclonal antibody specifically detects MBP in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| MBP | |
| Monoclonal | |
| Alexa Fluor 350 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Recombinant fragment (around aa 150-250) of human MBP (exact sequence is proprietary) (Uniprot: P02686) | |
| 100 μg | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
| 4155 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin), Protein Array | |
| MBP/4277R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction