missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MBTD1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582778

Product Code. 16379734

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TCWGDISENV RVEVPNTDCS LPTKVFWIAG IVKLAGYNAL LRYEGFENDS GLDFWCNICG SDIHPVGWCA ASGKPL Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility. Specifically binds to monomethylated and dimethylated 'Lys-20' on histone H4. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MBTD1
Polyclonal
Unconjugated
Mbtd1
AA408199; AI194990; hemp; im:7159307; mbt domain containing 1; MBT domain-containing protein 1; MBT domain-containing protein 1; LOW QUALITY PROTEIN: MBT domain-containing protein 1; MBTD1; SA49P01
Rabbit
Antigen affinity chromatography
RUO
54799
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q05BQ5
Mbtd1
Recombinant protein corresponding to Human MBTD1. Recombinant protein control fragment (Product #RP-93227).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.