Learn More
MBTD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26098-1-AP-20UL
 View more versions of this product
Product Code. 16824625
  • € 145.00 / 20µL
Description
MBDT1 is a putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility. MBDT1 specifically binds to monomethylated and dimethylated ′Lys-20′ on histone H4.
Specifications
Specifications
MBTD1
Polyclonal
Unconjugated
MBTD1
mbt domain containing 1, MBTD1, SA49P01
Rabbit
Antigen Affinity Chromatography
RUO
103537, 54799
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q05BQ5, Q6P5G3
MBTD1
MBTD1 Fusion Protein Ag22059
20 μL
Primary
Human, Mouse