missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBTPS1 Monoclonal Antibody (2F10E12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550278
This item is not returnable.
View return policy
Description
MBTPS1 Monoclonal Antibody for Western Blot, Flow, ELISA
This gene encodes a member of the subtilisin-like proprotein convertase family, which includes proteases that process protein and peptide precursors trafficking through regulated or constitutive branches of the secretory pathway. The encoded protein undergoes an initial autocatalytic processing event in the ER to generate a heterodimer which exits the ER and sorts to the cis/medial-Golgi where a second autocatalytic event takes place and the catalytic activity is acquired. It encodes a type 1 membrane bound protease which is ubiquitously expressed and regulates cholesterol or lipid homeostasis via cleavage of substrates at non-basic residues. Mutations in this gene may be associated with lysosomal dysfunction.
Specifications
| MBTPS1 | |
| Monoclonal | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q14703 | |
| MBTPS1 | |
| Recombinant Human Membrane-bound transcription factor site-1 protease protein (187-400aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 2F10E12 | |
| Unconjugated | |
| MBTPS1 | |
| 0610038M03Rik; AV003995; endopeptidase S1P; KIAA0091; MBTPS1; membrane bound transcription factor peptidase, site 1; membrane-bound transcription factor peptidase, site 1; membrane-bound transcription factor protease, site 1; membrane-bound transcription factor site-1 protease; mKIAA0091; PCSK8; proprotein convertase subtilisin/kexin type 8; S1p; S1P endopeptidase; site-1 protease; Ski1; SKI-1; Sterol-regulated luminal protease; subtilisin kexin isozyme 1; subtilisin/kexin isozyme 1; subtilisin/kexin isozyme SKI-1; subtilisin/kexin isozyme-1; Subtilisin/kexin-isozyme 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8720 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction