missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MC 1742

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
MC 1742 is a potent class I and IIb HDAC inhibitor (IC50 values are 7, 20, 40, 100, 110 and 610 nM for HDAC6, HDAC3, HDAC10, HDAC1, HDAC2 and HDAC8, respectively). Suppresses proliferation and induces apoptosis of sarcoma cancer stem cells (CSCs) at concentrations >500 nM. Also induces osteogenesis in sarcoma CSCs at concentrations of 25 - 500 nM .
Specifications
Specifications
| Chemical Name or Material | 5-[(4-[1,1′-Biphenyl]-4-yl-1,6-dihydro-6-oxo-2-pyrimidinyl)thio]-N-hydroxypentanamide |
| CAS | 1776116-74-5 |
| Quantity | 10 mg |
| Target | Histone Deacetylase Inhibitors |
| Molecular Formula | C21H21N3O3S |
| Formula Weight | 395.47 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction