missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MC2R Polyclonal Antibody
GREENER_CHOICE

Product Code. 17187833
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17187833 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17187833 Supplier Invitrogen™ Supplier No. PA5109906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MKHIINSYEN INNTARNNSD CPRVVLPEEI FFTISIVGVL ENLIVLLAVF KNKNLQAPMY FFICSLAISD MLGSLYKILE NILIILRNMG YLKPRGSFET.

MC2R encodes one member of the five-member G-protein associated melanocortin receptor family. Melanocortins are peptides derived from pro-opiomelanocortin. MC2R is selectively activated by adrenocorticotropic hormone, whereas the other four melanocortin receptors recognize a variety of melanocortin ligands. Mutations in MC2R can result in familial glucocorticoid deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MC2R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MC2R
Gene Accession No. Q01718, Q64326
Gene Alias ACTH receptor; ACTHR; ACTH-R; Adrenocorticotropic hormone receptor; adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2R; MC2-R; melanocortin 2 receptor; melanocortin 2 receptor (adrenocorticotropic hormone); melanocortin receptor 2; MGC125798
Gene Symbols MC2R
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human MC2R (NP_000520.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17200, 4158
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.