Learn More
Invitrogen™ MC5R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533930
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Monkey (100%); Marmoset, Mouse, Hamster, Panda (95%); Rabbit (89%); Rat, Dog, Bat, Opossum (84%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
MC5R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH), and activates adenylate cyclase. This receptor controls multiple functions in the brain and in peripheral tissues, such as thermoregulation, immunomodulation, appetite control, and sexual behavior. Melanocortin 5 receptor has been reported to be expressed in a variety of tissues, especially in pituitary, exocrine glands, reproductive organs, lung, kidney, and esophagus. No human ESTs have been isolated.
Specifications
| MC5R | |
| Polyclonal | |
| Unconjugated | |
| MC5R | |
| H MC2; MC2; MC-2; MC5; MC5 receptor; MC5R; MC5-R; melanocortin 5 receptor; melanocortin receptor 5; melanocortin-5 receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P33032 | |
| MC5R | |
| Synthetic 19 amino acid peptide from internal region of human MC5 Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.