missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MC5R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102100
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MC5R.
MC5R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH), and activates adenylate cyclase. This receptor controls multiple functions in the brain and in peripheral tissues, such as thermoregulation, immunomodulation, appetite control, and sexual behavior. Melanocortin 5 receptor has been reported to be expressed in a variety of tissues, especially in pituitary, exocrine glands, reproductive organs, lung, kidney, and esophagus. No human ESTs have been isolated.
Specifications
| MC5R | |
| Polyclonal | |
| Unconjugated | |
| MC5R | |
| H MC2; MC2; MC-2; MC5; MC5 receptor; MC5R; MC5-R; melanocortin 5 receptor; melanocortin receptor 5; melanocortin-5 receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17203, 25726, 4161 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P33032, P35345, P41149 | |
| MC5R | |
| A synthesized peptide derived from human MC5R(Accession P33032), corresponding to amino acid residues S275-D325. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction