missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM Monoclonal Antibody (OTI5C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803548S
This item is not returnable.
View return policy
Description
MCAM Monoclonal antibody specifically detects MCAM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MCAM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NYY1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 50604 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C4 | |
| Unconjugated | |
| Il20 | |
| 1-gicerin; AV025631; CD146; CD149; Cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; HEMCAM; l-gicerin; l-gicerin precursor; l-gicerin protein; LOW QUALITY PROTEIN: cell surface glycoprotein MUC18; MCAM; melanoma adhesion molecule; melanoma cell adhesion molecule; Melanoma-associated antigen A32; melanoma-associated antigen MUC18; Muc18; mucin-18; s-endo; S-endo 1 endothelial-associated antigen; s-gicerin; s-gicerin precursor | |
| Human recombinant protein fragment corresponding to amino acids 25-176 of human IL20 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction