missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAT Antibody (OTI2B7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72596CL1
This item is not returnable.
View return policy
Description
MCAT Monoclonal antibody specifically detects MCAT in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| MCAT | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.3.1.39, fabD, FASN2C, malonyl CoA:ACP acyltransferase (mitochondrial), malonyl-CoA:acyl carrier protein transacylase, mitochondrial, malonyl-CoA-acyl carrier protein transacylase, mitochondrial, MCT[Acyl-carrier-protein] malonyltransferase, Mitochondrial malonyltransferase, MTMGC47838, NET62 | |
| Full length human recombinant protein of human MCAT(NP_775738) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2B7 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 27349 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction