missing translation for 'onlineSavingsMsg'
Learn More

MCCC1, Mouse anti-Human, Clone: 2G8, Abnova™

Product Code. 16108876
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16108876 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16108876 Supplier Abnova Supplier No. H00056922M01.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence: FTLQAHDQFSPFSSSSGRRLNISYTRNMTLKDGKNNVAIAVTYNHDGSYSMQIEDKTFQVLGNLYSEGDCTYLKCSVNGVASKAKLIILENTIYLFSKEG

Specifications

Antigen MCCC1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
Gene Accession No. NM_020166
Gene Alias DKFZp686B20267/FLJ25545/MCC-B/MCCA
Gene Symbols MCCC1
Host Species Mouse
Immunogen MCCC1 (NP_064551, 526 a.a. ∼ 625 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56922
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.