missing translation for 'onlineSavingsMsg'
Learn More

MCCC2 Antibody [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30489941 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30489941 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30489941 Supplier Novus Biologicals Supplier No. NBP335509V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MCCC2 Polyclonal antibody specifically detects MCCC2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCCC2
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta, MCCase subunit beta, MCCBEC 6.4.1.4, methylcrotonoyl-CoA carboxylase 2 (beta), methylcrotonoyl-Coenzyme A carboxylase 2 (beta), mitochondrial, non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 284-563 of human MCCC2 (NP_071415.1).,, Sequence:, LHLTRKVVRNLNYQKKLDVTIEPSEEPLFPADELYGIVGANLKRSFDVREVIARIVDGSRFTEFKAFYGDTLVTGFARIFGYPVGIVGNNGVLFSESAKKGTHFVQLCCQRNIPLLFLQNITGFMVGREYEAEGIAKDGAKMVAAVACAQVPKITLIIGGSYGAGNYGMCGRAYSPRFLYIWPNARISVMGGEQAANVLATITKDQRAREGKQFSSADEAALKEPIIKKFEEEGNPYYSSARVWDDGIIDPADTRLVLGLSFSAALNAPIEKTDFGIFRM
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64087
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.