missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCCC2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97284JF646
This item is not returnable.
View return policy
Description
MCCC2 Polyclonal specifically detects MCCC2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MCCC2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 64087 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta, MCCase subunit beta, MCCBEC 6.4.1.4, methylcrotonoyl-CoA carboxylase 2 (beta), methylcrotonoyl-Coenzyme A carboxylase 2 (beta), mitochondrial, non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase | |
| Produced in rabbits immunized with E. coli-derived Human MCCC2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction