missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCEE Antibody (OTI1B5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72597AF700
This item is not returnable.
View return policy
Description
MCEE Monoclonal specifically detects MCEE in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MCEE | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MCEE | |
| Full length recombinant protein of human MCEE (NP_115990) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1B5 | |
| Western Blot, Immunohistochemistry | |
| DL-methylmalonyl-CoA racemase, EC 5.1.99.1, GLOD2, glyoxalase domain containing 2, methylmalonyl CoA epimerase, methylmalonyl-CoA epimerase, mitochondrial | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 84693.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction