missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCEE Monoclonal Antibody (OTI3B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808576S
This item is not returnable.
View return policy
Description
MCEE Monoclonal antibody specifically detects MCEE in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MCEE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96PE7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84693 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B11 | |
| Unconjugated | |
| MCEE | |
| 1110007A04Rik; DL-methylmalonyl-CoA racemase; GLOD2; glyoxalase domain containing 2; Mcee; methylmalonyl CoA epimerase; methylmalonyl-CoA epimerase; methylmalonyl-CoA epimerase, mitochondrial | |
| Full length human recombinant protein of human MCEE produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction