Learn More
MCEE Mouse anti-Human, Clone: OTI4A6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808533
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.Specifications
| MCEE | |
| Monoclonal | |
| Unconjugated | |
| MCEE | |
| GLOD2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84693 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4A6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96PE7 | |
| MCEE | |
| Full length human recombit protein of human MCEE produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.