Learn More
MCF2 Mouse anti-Human, Clone: OTI3E4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812542
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Guanine nucleotide exchange factor (GEF) that modulates the Rho family of GTPases. Promotes the conversion of some member of the Rho family GTPase from the GDP-bound to the GTP-bound form. Isoform 1 exhibits no activity toward RHOA, RAC1 or CDC42. Isoform 2 exhibits decreased GEF activity toward CDC42. Isoform 3 exhibits a weak but significant activity toward RAC1 and CDC42. Isoform 4 exhibits significant activity toward RHOA and CDC42. The truncated DBL oncogene is active toward RHOA, RAC1 and CDC42.Specifications
| MCF2 | |
| Monoclonal | |
| Unconjugated | |
| MCF2 | |
| ARHGEF21, DBL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4168 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3E4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P10911 | |
| MCF2 | |
| Human recombit protein fragment corresponding to amino acids 872-1001 of human MCF2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.