missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCFD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15508512
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15508512 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508512 Supplier Invitrogen™ Supplier No. PA548016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Multiple coagulation factor deficiency protein 2 (MCFD2) is localized in the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) through a direct, calcium-dependent interaction with LMAN1. The MCFD2-LMAN1 complex forms a specific cargo receptor for the transport of selected proteins from the endoplasmic reticulum to the Golgi apparatus. Mutations in the MCFD2 gene may cause of factor V and factor VIII combined deficiency (F5F8D). F5F8D is an autosomal recessive human bleeding disorder characterized by the reduction of both clotting proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCFD2
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Mcfd2
Gene Accession No. Q8K5B2
Gene Alias 1810021C21Rik; F5F8D; F5F8D2; LMAN1IP; MCFD2; multiple coagulation factor deficiency 2; multiple coagulation factor deficiency protein 2; Multiple coagulation factor deficiency protein 2 homolog; neural stem cell derived neuronal survival protein; neural stem cell-derived neuronal survival protein; Sdnsf; stem cell derived neuronal survival protein
Gene Symbols Mcfd2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SDNSF/MCFD2 His27-Gln145.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 193813
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.