missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody (1C51), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-96752AF700
This item is not returnable.
View return policy
Description
mCherry Monoclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunoassay.
Specifications
| mCherry | |
| Monoclonal | |
| Alexa Fluor 700 | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| This mCherry Antibody (1C51) was developed against recombinant full-length mCherry purified from E. coli. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Non-species specific, Mouse, Rat, Drosophila, Insect | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoassay | |
| 1C51 | |
| Western Blot, Immunohistochemistry, Knockout Validated | |
| Mouse | |
| 27 kDa | |
| 0.1 mL | |
| Cellular Markers, Fusion Proteins | |
| This mCherry Antibody (1C51) does not cross react with GFP. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction