missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody, Novus Biologicals™
Goat Polyclonal Antibody
€ 339.00 - € 572.00
Specifications
| Antigen | mCherry |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, Immunocytochemistry/ Immunofluorescence 1:1000 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697597
|
Novus Biologicals
NBP3-05558-100ul |
100 μg |
€ 572.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691428
|
Novus Biologicals
NBP3-05558-50ul |
50 μg |
€ 339.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mCherry Polyclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| mCherry | |
| Western Blot 1:2000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Goat | |
| Cellular Markers, Fusion Proteins | |
| 50% PBS, 50% glycerol | |
| This mCherry Antibody was developed against full length recombinant mCherry protein | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title