missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mCherry Monoclonal Antibody (5A6)

Mouse Monoclonal Antibody

€ 1704.00

Specifications

Antigen mCherry
Clone 5A6
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17856827
View Documents
Invitrogen™
MA547341
500 μL
€ 1704.00
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17886627
View Documents
Invitrogen™
MA547344
100 μL
€ 1704.00
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

mCherry Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
Specifications

Specifications

mCherry
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 5mM sodium azide
Full length recombinant mCherry protein expressed in and purified from E. coli.
Antibody
5A6
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG1 κ
Chemical
mCherry
Primary
Protein G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.