missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mCherry Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143590
This item is not returnable.
View return policy
Description
mCherry Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
Specifications
| mCherry | |
| Polyclonal | |
| Unconjugated | |
| mCherry | |
| Full length recombinant mCherry protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction