missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mCherry Recombinant Rat Monoclonal Antibody (16D7), HRP, Invitrogen™

Product Code. 30019225
Change view
Click to view available options
Quantity:
20 μL
50 μL
Unit Size:
20µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019225 50 μL 50µg
30019534 20 μL 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30019225 Supplier Invitrogen™ Supplier No. 740003THRP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Recombinant Monoclonal Antibody

mCherry Recombinant Monoclonal antibody specifically detects mCherry in Tag samples. It is validated for Western Blot

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Western Blot
Classification Recombinant Monoclonal
Clone 16D7
Concentration 1.0 mg/mL
Conjugate HRP
Formulation PBS with proprietary stabilizer and proprietary preservative; pH 7.4
Gene Alias mCherry
Host Species Rat
Immunogen full-length protein mCherry.
Purification Method Affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.