missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCHR1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15634635
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15634635 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15634635 Supplier Invitrogen™ Supplier No. PA550705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SLC1 / MCHR1 protein.

MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCHR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Mchr1
Gene Accession No. P97639, Q8JZL2, Q99705
Gene Alias AW049955; G protein coupled receptor 24; G protein-coupled receptor 24; GPCR24; GPR24; Gpr24-9 protein coupled receptor; G-protein coupled receptor 24; MCH 1 receptor; MCH receptor 1; MCH1; Mch1r; MCH-1R; MCHR; MCHR1; MCH-R1; MCHR-1; melanin concentrating hormone receptor 1; melanin-concentrating hormone receptor 1; SLC1; SLC-1; Somatostatin receptor-like protein
Gene Symbols Mchr1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human melanin-concentrating hormone receptor 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207911, 2847, 83567
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.