missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mcl-1 Antibody (OTI10F6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72598FR
This item is not returnable.
View return policy
Description
Mcl-1 Monoclonal specifically detects Mcl-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Mcl-1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| MCL1 | |
| Full length recombinant protein of human Mcl-1 (NP_068779) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI10F6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| bcl2-L-3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Signal Transduction, Tumor Suppressors | |
| 4170.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction