Learn More
Mcl-1 Antibody (S04-3A4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19546-50ul
 Afficher plus de versions de ce produit
Code produit. 18614055
  • € 421.00 / 50µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Mcl-1 Monoclonal antibody specifically detects Mcl-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Mcl-1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunofluorescence
S04-3A4
Unconjugated
bcl2-L-3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM
A synthetic peptide of human Mcl-1 (Uniprot # Q07820)
50 μg
Apoptosis, Cancer, Signal Transduction, Tumor Suppressors
4170
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.