Learn More
MCL1 Mouse anti-Human, Clone: OTI3A12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500983
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MCL1 belongs to the Bcl-2 family. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. The longer gene product (isoform 1) enhances cell survival by inhibiting apoptosis while the alternatively spliced shorter gene product (isoform 2) promotes apoptosis and is death-inducing.Specifications
| MCL1 | |
| Monoclonal | |
| Unconjugated | |
| MCL1 | |
| BCL2L3, EAT, MCL1-ES, MCL1L, MCL1S, Mcl-1, TM, bcl2-L-3, mcl1/EAT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4170 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q07820 | |
| MCL1 | |
| Full length human recombit protein of human MCL1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.