missing translation for 'onlineSavingsMsg'
Learn More

MCL1 Rabbit Recombinant Antibody, Proteintech

Rabbit Recombinant Antibody

€ 149.00 - € 439.00

Specifications

Antigen MCL1
Clone 1C7
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236297
View Documents
Proteintech
82695-2-RR-20UL
20 μL
€ 149.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235127
View Documents
Proteintech
82695-2-RR-100UL
100 μL
€ 439.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MCL1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MCL1. This antibody recognizes human, mouse, rat antigen. The MCL1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.
Specifications

Specifications

MCL1
1000 μg/mL
Recombinant Monoclonal
Liquid
RUO
4170
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
1C7
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Unconjugated
Rabbit
Human, Mouse, Rat
BCL2L3; EAT; MCL1L; MCL1S; MGC104264; MGC1839; Mcl-1; TM
IgG
Protein A purification
Recombinant Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.