missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM10 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-253UV
This item is not returnable.
View return policy
Description
MCM10 Polyclonal antibody specifically detects MCM10 in Human samples. It is validated for Western Blot
Specifications
| MCM10 | |
| Polyclonal | |
| Western Blot | |
| CNA43homolog of yeast MCM10, DNA43, hsMCM10, MCM10 minichromosome maintenance deficient 10, MCM10 minichromosome maintenance deficient 10 (S. cerevisiae), MGC126776, minichromosome maintenance complex component 10, PRO2249, protein MCM10 homolog | |
| The immunogen recognized by this antibody maps to a region between residues 600 and 650 of human Minichromosome Maintenance 10 using the numbering given in entry NP_060988.3 (GeneID 55388). | |
| 0.1 mL | |
| DNA Repair | |
| 55388 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction