missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM2 Antibody, DyLight 350, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-244UV
This item is not returnable.
View return policy
Description
MCM2 Polyclonal antibody specifically detects MCM2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MCM2 | |
| Polyclonal | |
| Western Blot | |
| BM28MGC10606, CCNL 1, CCNL1, CDCL1mitotin, EC 3.6.4.12, KIAA0030MITOTIN, MCM2 minichromosome maintenance deficient 2, mitotin (S. cerevisiae), minichromosome maintenance complex component 2, minichromosome maintenance deficient (S. cerevisiae) 2 (mitotin), minichromosome maintenance deficient 2 (mitotin) | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Minichromosomal Maintenance Deficient 2 using the numbering given in entry NP_0045117.2 (GeneID 4171). | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| 4171 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction