missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM2 Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805472S
This item is not returnable.
View return policy
Description
MCM2 Monoclonal antibody specifically detects MCM2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MCM2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06401 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5241 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G4 | |
| Unconjugated | |
| PGR | |
| AA959861; AW476101; BM28; CCNL1; cdc19; CDCL1; cell devision cycle-like 1; cyclin-like 1; D3S3194; DFNA70; DNA replication licensing factor MCM2; Kiaa0030; MCM DNA helicase complex subunit MCM2; Mcm2; Mcmd2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance complex component 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; minichromosome maintenance deficient 2 mitotin (S. cerevisiae); Minichromosome maintenance protein 2; Minichromosome maintenance protein 2 homolog; MITOTIN; mKIAA0030; Nuclear protein BM28; YBL023C; YBL0438 | |
| Human recombinant protein fragment corresponding to amino acids 1-298 of human PGR produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction