missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM2 Monoclonal Antibody (OTI8A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805566
This item is not returnable.
View return policy
Description
MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).Specifications
| MCM2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49736 | |
| MCM2 | |
| Full length human recombit protein of human MCM2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8A11 | |
| Unconjugated | |
| MCM2 | |
| BM28, CCNL1, CDCL1, D3S3194, DFNA70, MITOTIN, cdc19 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4171 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction