missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM3 Antibody, CoraFluor™ 1, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-249CL1
This item is not returnable.
View return policy
Description
MCM3 Polyclonal antibody specifically detects MCM3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
Specifications
| MCM3 | |
| Polyclonal | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| CoraFluor 1 | |
| cervical cancer proto-oncogene 5, DNA polymerase alpha holoenzyme-associated protein P1, DNA replication factor MCM3, DNA replication licensing factor MCM3, EC 3.6.4.12, HCC5, hRlf beta subunit, MCM3 minichromosome maintenance deficient 3, MCM3 minichromosome maintenance deficient 3 (S. cerevisiae), MGC1157, minichromosome maintenance complex component 3, minichromosome maintenance deficient (S. cerevisiae) 3, minichromosome maintenance deficient 3, P1.h, p102, P1-MCM3, replication licensing factor, beta subunit, RLF subunit beta, RLFB | |
| The immunogen recognized by this antibody maps to a region between residues 500 and 550 of human Minichromosome Maintenance 3 using the numbering given in entry NP_002379.2 (GeneID 4172). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4172 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction