missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCM4 Recombinant Rabbit Monoclonal Antibody (23GB2495), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | MCM4 |
|---|---|
| Clone | 23GB2495 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30087459
|
Invitrogen™
MA552516 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089424
|
Invitrogen™
MA552517 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCM4 Recombinant Monoclonal antibody specifically detects MCM4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MCM4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P33991, P49717 | |
| 17217, 4173 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2495 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 19G; AI325074; AU045576; cb1025; Cdc21; CDC21 homolog; CDC54; DNA replication licensing factor MCM4; fc12c09; fj85g09; hCdc21; hm:zeh1616; homolog of S. pombe cell devision cycle 21; mcdc21; MCM4; MCM4 minichromosome maintenance deficient 4; MCM4 minichromosome maintenance deficient 4, mitotin; mcm4 protein; Mcmd4; mini chromosome maintenance deficient 4 homolog; minichromosome maintenance complex component 4; minichromosome maintenance deficient 4; minichromosome maintenance deficient 4 homolog; minichromosome maintenance deficient 4 homolog (S. cerevisiae); mKIAA4003; NKCD; NKGCD; P1-CDC21; Unknown (protein for MGC:140484); wu:fc12c09; wu:fj85g09; zeh1616 | |
| A synthesized peptide derived from human MCM4 (820-863AA). | |
| Primary | |
| -20°C | |
| Mcm4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title