missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM5 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79778FR
This item is not returnable.
View return policy
Description
MCM5 Polyclonal antibody specifically detects MCM5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MCM5 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| CDC46 homolog, CDC46DNA replication licensing factor MCM5, EC 3.6.4.12, MCM5 minichromosome maintenance deficient 5, cell division cycle 46, MCM5 minichromosome maintenance deficient 5, cell division cycle 46 (S.cerevisiae), MGC5315, minichromosome maintenance complex component 5, minichromosome maintenance deficient (S. cerevisiae) 5 (cell division cycle 46), minichromosome maintenance deficient 5 (cell division cycle 46), P1-CDC46 | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Minichromosome Maintenance 5 using the numbering given in entry NP_006730.2 (GeneID 4174). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4174 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction