missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCM5 Recombinant Rabbit Monoclonal Antibody (4P1Q5)

MCM5 Recombinant Rabbit Monoclonal Antibody (ARC1239)
Brand: Invitrogen™ MA535619
This item is not returnable.
View return policy
Description
MCM5 Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
Specifications
| MCM5 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P33992, P49718 | |
| MCM5 | |
| A synthetic peptide corresponding to a sequence within amino acids 635-734 of human MCM5 (P33992). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 4P1Q5 | |
| Unconjugated | |
| MCM5 | |
| AA617332; AI324988; AL033333; Cdc46; CDC46 homolog; DNA replication licensing factor MCM5; MCM5; MCM5 minichromosome maintenance deficient 5, cell division cycle 46; Mcmd5; MGC5315; minichromosome maintenance complex component 5; minichromosome maintenance deficient 5 (cell division cycle 46); minichromosome maintenance deficient 5, cell division cycle 46; minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae); P1-CDC46 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17218, 291885, 4174 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction