Learn More
MCM6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-13347-100UL
 View more versions of this product
Product Code. 18018745
  • € 450.00 / 100µL
Description
The MCM6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MCM6. This antibody recognizes human, mouse, rat antigen. The MCM6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
MCM6
1000 μg/mL
MCG40308, MCM6, Mis5, P105MCM
100 μL
4175
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa