Learn More
MCM6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-13347-100UL
Description
The MCM6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MCM6. This antibody recognizes human, mouse, rat antigen. The MCM6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| MCM6 | |
| 1000 μg/mL | |
| MCG40308, MCM6, Mis5, P105MCM | |
| 100 μL | |
| 4175 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |