missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCM6 Recombinant Rabbit Monoclonal Antibody (JE65-68)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544691
This item is not returnable.
View return policy
Description
MCM6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
MCM6 may be involved in the control of a single round of DNA replication during S phase. It binds to chromatin during G1 and detaches from it during S phase as if it licenses the chromatin to replicate.
Specifications
| MCM6 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P97311, Q14566, Q62724 | |
| Mcm6 | |
| Recombinant protein within human MCM6 aa 572-821/821. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE65-68 | |
| Unconjugated | |
| Mcm6 | |
| ASP-l1; D1Wsu22e; DNA replication licensing factor MCM6; intestinal DNA replication protein; MCG40308; Mcm6; MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); Mcmd6; mini chromosome maintenance deficient 6; minichromosome maintenance complex component 6; minichromosome maintenance deficient (mis5, S. pombe) 6; minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S. cerevisiae); Mis5; Mis5 homolog; p105MCM | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17219, 29685, 4175 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction