Learn More
MCM7 Antibody (2068B), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB9217MFV450
Product Code. 18690910
  • € 608.00 / 0.10mL
Description
MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry, Flow Cytometry
CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
RUO
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry
E. coli-derived recombinant human MCM7, Gly188-Ala328, Accession # P33993